1-(2,3-Dichlorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65989-1G 65989-500MG 65989-5G
77472-528EA 594.31 USD
77472-528 77485-152 77495-716
1-(2,3-Dichlorophenyl)-2,2-difluoroethanone ≥95%
1-(2,3-Dichlorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₄Cl₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD26045120
CAS Number: 1565371-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR