1-(2-Chloro-5-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65967-1G 65967-500MG 65967-5G 65967-25G
77472-524EA 416.12 USD
77472-524 77485-148 77495-712 77504-986
1-(2-Chloro-5-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(2-Chloro-5-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD28105302
CAS Number: 1597054-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR