2-Ethyl-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

65912-1G 65912-500MG 65912-5G
77472-520EA 543.84 USD
77472-520 77485-144 77495-706
2-Ethyl-[1,1'-biphenyl]-4-carbaldehyde ≥95%
2-Ethyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O
MW: 210.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762322
CAS Number: 2807469-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR