1-(3,4-Diethoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65795-1G 65795-500MG 65795-5G 65795-25G
77472-516EA 415.09 USD
77472-516 77485-140 77495-702 77504-980
1-(3,4-Diethoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3,4-Diethoxyphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₂H₁₃F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD07380820
CAS Number: 1408457-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR