1-(2-Chloro-3-fluoropyridin-4-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65767-1G 65767-500MG 65767-5G 65767-25G
77472-512EA 415.09 USD
77472-512 77485-136 77495-698 77504-978
1-(2-Chloro-3-fluoropyridin-4-yl)-2,2-difluoroethanone ≥95%
1-(2-Chloro-3-fluoropyridin-4-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₇H₃ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD34762303
CAS Number: 2901106-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR