2,2-dDifluoro-1-(3-fluoropyridin-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65735-1G 65735-500MG 65735-5G
77472-508EA 640.66 USD
77472-508 77485-132 77495-694
2,2-dDifluoro-1-(3-fluoropyridin-4-yl)ethanone ≥95%
2,2-dDifluoro-1-(3-fluoropyridin-4-yl)ethanone

Precaución: For research use only

Formula: C₇H₄F₃NO
MW: 175.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34814422
CAS Number: 1565418-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR