2,2,2-Trifluoro-1-(4-morpholinophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65694-1G 65694-500MG 65694-5G
77472-504EA 442.9 USD
77472-504 77485-128 77495-690
2,2,2-Trifluoro-1-(4-morpholinophenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-morpholinophenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₂F₃NO₂
MW: 259.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11556207
CAS Number: 70783-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR