5-Chloro-4-ethoxy-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65596-1G 65596-500MG 65596-5G 65596-25G
77472-500EA 415.09 USD
77472-500 77485-124 77495-686 77504-970
5-Chloro-4-ethoxy-2-methoxybenzaldehyde ≥97%
5-Chloro-4-ethoxy-2-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762271
CAS Number: 2385274-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR