1-(3,6-Difluoro-2-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65485-1G 65485-500MG 65485-5G
77472-496EA 696.28 USD
77472-496 77485-120 77495-682
1-(3,6-Difluoro-2-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3,6-Difluoro-2-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₉H₅F₅O₂
MW: 240.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922641
CAS Number: 1208077-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR