5-Bromo-4-methoxynicotinaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
65442-1G
65442-500MG
65442-5G
65442-25G
77472-492EA
189.52
USD
77472-492
77485-116
77495-678
77504-966
5-Bromo-4-methoxynicotinaldehyde ≥97%
5-Bromo-4-methoxynicotinaldehyde
Precaución: For research use only
Formula:
C₇H₆BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28740848 CAS Number: 1289161-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...