Trimethyl(2,3,5,6-tetrafluorophenyl)silane ≥95%

Supplier: AOBChem USA

Warning

65302-1G 65302-500MG 65302-5G
77472-488EA 546.93 USD
77472-488 77485-112 77495-672
Trimethyl(2,3,5,6-tetrafluorophenyl)silane ≥95%
Trimethyl(2,3,5,6-tetrafluorophenyl)silane

Precaución: For research use only

Formula: C₉H₁₀F₄Si
MW: 222.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092631
CAS Number: 20083-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR