2-(5-Bromo-4-chloro-2-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

65289-1G 65289-500MG 65289-5G 65289-25G
77472-484EA 415.09 USD
77472-484 77485-108 77495-668 77504-958
2-(5-Bromo-4-chloro-2-fluorophenyl)-1,3-dioxolane ≥97%
2-(5-Bromo-4-chloro-2-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇BrClFO₂
MW: 281.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762179
CAS Number: 2807467-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR