(5-Bromo-2,3,4-trifluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

65235-1G 65235-500MG 65235-5G 65235-25G
77472-480EA 129.78 USD
77472-480 77485-104 77495-664 77504-954
(5-Bromo-2,3,4-trifluorophenyl)trimethylsilane ≥95%
(5-Bromo-2,3,4-trifluorophenyl)trimethylsilane

Precaución: For research use only

Formula: C₉H₁₀BrF₃Si
MW: 283.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28953351
CAS Number: 1936228-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR