2,2,2-Trifluoro-1-(2-fluoro-6-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65217-1G 65217-500MG 65217-5G 65217-25G
77472-476EA 415.09 USD
77472-476 77485-100 77495-660 77504-950
2,2,2-Trifluoro-1-(2-fluoro-6-(trifluoromethyl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-fluoro-6-(trifluoromethyl)phenyl)ethanone

Precaución: For research use only

Formula: C₉H₃F₇O
MW: 260.1083 g/mol
Storage Temperature: Ambient
CAS Number: 1519776-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR