1-(6-Bromo-2-chloro-3-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65199-1G 65199-500MG 65199-5G 65199-25G
77472-472EA 415.09 USD
77472-472 77485-096 77495-656 77504-948
1-(6-Bromo-2-chloro-3-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(6-Bromo-2-chloro-3-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O₂
MW: 317.4870 g/mol
Storage Temperature: Ambient
CAS Number: 3002529-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR