2,2,2-trifluoro-1-(3-(methoxymethoxy)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65121-1G 65121-500MG 65121-5G 65121-25G
77472-464EA 415.09 USD
77472-464 77485-088 77495-648 77504-944
2,2,2-trifluoro-1-(3-(methoxymethoxy)phenyl)ethanone ≥95%
2,2,2-trifluoro-1-(3-(methoxymethoxy)phenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₉F₃O₃
MW: 234.1718 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762103
CAS Number: 113787-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR