3',4'-Dimethoxy-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

65115-1G 65115-5G 65115-25G
77472-460EA 60.77 USD
77472-460 77495-644 77504-942
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone ≥97%
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone

Precaución: For research use only

Formula: C₁₀H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD01319991
CAS Number: 300374-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR