1-Bromo-5-fluoro-2-(methoxymethoxy)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

65051-1G 65051-500MG 65051-5G 65051-25G
77472-452EA 415.09 USD
77472-452 77485-078 77495-636 77504-936
1-Bromo-5-fluoro-2-(methoxymethoxy)-4-methylbenzene ≥97%
1-Bromo-5-fluoro-2-(methoxymethoxy)-4-methylbenzene

Precaución: For research use only

Formula: C₉H₁₀BrFO₂
MW: 249.0769 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762077
CAS Number: 162269-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR