6-(Cyclopropylmethoxy)-5-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

64958-1G 64958-500MG 64958-5G
77472-448EA 597.4 USD
77472-448 77485-074 77495-632
6-(Cyclopropylmethoxy)-5-methylnicotinaldehyde ≥97%
6-(Cyclopropylmethoxy)-5-methylnicotinaldehyde

Precaución: For research use only

Formula: C₁₁H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD23952658
CAS Number: 1289127-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR