2-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

64871-1G 64871-500MG 64871-5G 64871-25G
77472-444EA 415.09 USD
77472-444 77485-070 77495-628 77504-932
2-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde ≥95%
2-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₃BrO₃
MW: 321.1659 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762003
CAS Number: 2247294-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR