(2-Chloro-6-fluoro-4-methoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

64837-1G 64837-500MG 64837-5G 64837-25G
77472-440EA 111.24 USD
77472-440 77485-066 77495-624 77504-928
(2-Chloro-6-fluoro-4-methoxyphenyl)boronic acid ≥95%
(2-Chloro-6-fluoro-4-methoxyphenyl)boronic acid

Precaución: For research use only

Formula: C₇H₇BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD31640727
CAS Number: 2044684-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR