3,5-Dibromo-2-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

64800-1G 64800-500MG 64800-5G 64800-25G
77472-436EA 216.3 USD
77472-436 77485-062 77495-618 77504-922
3,5-Dibromo-2-isopropoxybenzaldehyde ≥95%
3,5-Dibromo-2-isopropoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02257453
CAS Number: 832673-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR