5-Chloro-3-fluoro-2-methoxy-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

64756-1G 64756-500MG 64756-5G 64756-25G
77472-432EA 140.08 USD
77472-432 77485-058 77495-614 77504-918
5-Chloro-3-fluoro-2-methoxy-4-(methylthio)pyridine ≥97%
5-Chloro-3-fluoro-2-methoxy-4-(methylthio)pyridine

Precaución: For research use only

Formula: C₇H₇ClFNOS
MW: 207.6529 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761957
CAS Number: 2807471-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR