3,5-Dichloro-2-methoxyisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

64729-1G 64729-500MG 64729-5G 64729-25G
77472-428EA 197.76 USD
77472-428 77485-054 77495-610 77504-914
3,5-Dichloro-2-methoxyisonicotinaldehyde ≥97%
3,5-Dichloro-2-methoxyisonicotinaldehyde

Precaución: For research use only

Formula: C₇H₅Cl₂NO₂
MW: 206.0261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31645820
CAS Number: 1884279-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR