4-(Dimethylamino)-2,6-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64722-1G 64722-500MG 64722-5G 64722-25G
77472-424EA 226.6 USD
77472-424 77485-050 77495-606 77504-910
4-(Dimethylamino)-2,6-difluorobenzoic acid ≥97%
4-(Dimethylamino)-2,6-difluorobenzoic acid

Precaución: For research use only

Formula: C₉H₉F₂NO₂
MW: 201.1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32140541
CAS Number: 1697868-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR