(5-Bromo-2-fluoro-3-isobutoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

64695-1G 64695-500MG 64695-5G
77472-420EA 486.16 USD
77472-420 77485-046 77495-602
(5-Bromo-2-fluoro-3-isobutoxyphenyl)(methyl)sulfane ≥95%
(5-Bromo-2-fluoro-3-isobutoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₄BrFOS
MW: 293.1956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761919
CAS Number: 2807446-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR