(2-Fluoro-6-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

64668-1G 64668-500MG 64668-5G 64668-25G
77472-416EA 175.1 USD
77472-416 77485-042 77495-598 77504-906
(2-Fluoro-6-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-6-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₉FS
MW: 156.2204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761901
CAS Number: 2807469-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR