4'-Methoxy-2-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64600-1G 64600-500MG 64600-5G 64600-25G
77472-412EA 415.09 USD
77472-412 77485-038 77495-594 77504-902
4'-Methoxy-2-nitro-1,1'-biphenyl ≥97%
4'-Methoxy-2-nitro-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₃H₁₁NO₃
MW: 229.2313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909442
CAS Number: 20013-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR