3-(4-Bromo-3-(methylthio)phenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

64590-1G 64590-500MG 64590-5G 64590-25G
77472-408EA 415.09 USD
77472-408 77485-034 77495-590 77504-898
3-(4-Bromo-3-(methylthio)phenyl)pyridine ≥97%
3-(4-Bromo-3-(methylthio)phenyl)pyridine

Precaución: For research use only

Formula: C₁₂H₁₀BrNS
MW: 280.1835 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761850
CAS Number: 2901101-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR