(2-(Pyrrolidin-1-yl)pyridin-4-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

64576-1G 64576-500MG 64576-5G 64576-25G
77472-404EA 335.78 USD
77472-404 77485-030 77495-586 77504-894
(2-(Pyrrolidin-1-yl)pyridin-4-yl)boronic acid ≥97%
(2-(Pyrrolidin-1-yl)pyridin-4-yl)boronic acid

Precaución: For research use only

Formula: C₉H₁₃BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD22566399
CAS Number: 2225155-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR