1-Chloro-3-fluoro-2-(methoxymethoxy)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

64512-1G 64512-500MG 64512-5G 64512-25G
77472-400EA 114.33 USD
77472-400 77485-026 77495-582 77504-890
1-Chloro-3-fluoro-2-(methoxymethoxy)-4-methylbenzene ≥97%
1-Chloro-3-fluoro-2-(methoxymethoxy)-4-methylbenzene

Precaución: For research use only

Formula: C₉H₁₀ClFO₂
MW: 204.6259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761808
CAS Number: 2807456-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR