5-Bromo-2-fluoro-3,4-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64437-1G 64437-500MG 64437-5G 64437-25G
77472-396EA 162.74 USD
77472-396 77485-022 77495-578 77504-886
5-Bromo-2-fluoro-3,4-dimethylbenzaldehyde ≥97%
5-Bromo-2-fluoro-3,4-dimethylbenzaldehyde

Precaución: For research use only

Formula: C₉H₈BrFO
MW: 231.0616 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761768
CAS Number: 2807471-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR