1-(2,3-Difluoropyridin-4-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

64393-1G 64393-500MG 64393-5G 64393-25G
77472-392EA 141.11 USD
77472-392 77485-018 77495-574 77504-884
1-(2,3-Difluoropyridin-4-yl)ethanone ≥97%
1-(2,3-Difluoropyridin-4-yl)ethanone

Precaución: For research use only

Formula: C₇H₅F₂NO
MW: 157.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31567993
CAS Number: 2169647-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR