1-(Benzo[b]thiophen-7-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

64219-1G 64219-500MG 64219-5G
77472-380EA 696.28 USD
77472-380 77485-006 77495-560
1-(Benzo[b]thiophen-7-yl)-2,2,2-trifluoroethanone ≥95%
1-(Benzo[b]thiophen-7-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₀H₅F₃OS
Storage Temperature: Ambient
CAS Number: 1514166-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR