2-(3-Ethyl-4-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

64190-1G 64190-500MG 64190-5G 64190-25G
77472-376EA 197.76 USD
77472-376 77485-002 77495-556 77504-870
2-(3-Ethyl-4-fluorophenyl)-1,3-dioxolane ≥97%
2-(3-Ethyl-4-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34761660
CAS Number: 2807469-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR