(2,5-Difluoropyridin-4-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

64170-1G 64170-500MG 64170-5G 64170-25G
77472-372EA 177.16 USD
77472-372 77484-998 77495-552 77504-866
(2,5-Difluoropyridin-4-yl)methanol ≥95%
(2,5-Difluoropyridin-4-yl)methanol

Precaución: For research use only

Formula: C₆H₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD12756645
CAS Number: 1227601-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR