2,2,2-Trifluoro-1-(5-fluoro-2-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
64086-1G
64086-500MG
64086-5G
64086-25G
77472-368EA
119.48
USD
77472-368
77484-994
77495-548
77504-862
2,2,2-Trifluoro-1-(5-fluoro-2-methoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(5-fluoro-2-methoxyphenyl)ethanone
Precaución: For research use only
Formula:
C₉H₆F₄O₂ MW: 222.1363 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07380760 CAS Number: 1256467-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...