(2-Fluoro-4-iodo-3-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63936-1G 63936-500MG 63936-5G
77472-364EA 914.64 USD
77472-364 77484-990 77495-544
(2-Fluoro-4-iodo-3-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-4-iodo-3-(methoxymethoxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀FIO₂S
Storage Temperature: Ambient
MDL Number: MFCD34761574
CAS Number: 2901066-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR