2-Fluoro-4-iodo-3-(methoxymethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63933-1G 63933-500MG 63933-5G 63933-25G
77472-360EA 415.09 USD
77472-360 77484-986 77495-542 77504-858
2-Fluoro-4-iodo-3-(methoxymethoxy)benzaldehyde ≥97%
2-Fluoro-4-iodo-3-(methoxymethoxy)benzaldehyde

Precaución: For research use only

Formula: C₉H₈FIO₃
Storage Temperature: Ambient
MDL Number: MFCD34761572
CAS Number: 2901082-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR