1-(2,6-Dichloro-3-methylphenyl)-2,2,2-trifluoroethanol ≥95%

Supplier: AOBChem USA

Warning

63925-1G 63925-500MG 63925-5G 63925-25G
77472-356EA 415.09 USD
77472-356 77484-982 77495-538 77504-854
1-(2,6-Dichloro-3-methylphenyl)-2,2,2-trifluoroethanol ≥95%
1-(2,6-Dichloro-3-methylphenyl)-2,2,2-trifluoroethanol

Precaución: For research use only

Formula: C₉H₇Cl₂F₃O
MW: 259.0524 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761566
CAS Number: 2918952-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR