2,2,2-Trifluoro-1-(2-methoxy-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63851-1G 63851-500MG 63851-5G
77472-348EA 692.16 USD
77472-348 77484-974 77495-530
2,2,2-Trifluoro-1-(2-methoxy-4-methylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-methoxy-4-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD13173387
CAS Number: 145965-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR