2',3'-Dimethyl-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

63756-1G 63756-500MG 63756-5G 63756-25G
77472-340EA 99.91 USD
77472-340 77484-966 77495-522 77504-840
2',3'-Dimethyl-2,2,2-trifluoroacetophenone ≥95%
2',3'-Dimethyl-2,2,2-trifluoroacetophenone

Precaución: For research use only

Formula: C₁₀H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD01319983
CAS Number: 886369-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR