4-(Benzyloxy)-3-(tert-butyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63716-1G 63716-500MG 63716-5G
77472-336EA 546.93 USD
77472-336 77484-962 77495-518
4-(Benzyloxy)-3-(tert-butyl)benzaldehyde ≥97%
4-(Benzyloxy)-3-(tert-butyl)benzaldehyde

Precaución: For research use only

Formula: C₁₈H₂₀O₂
MW: 268.3502 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19439976
CAS Number: 129357-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR