2,2,2-trifluoro-1-(2-(methoxymethoxy)-4,5-dimethylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63647-1G 63647-500MG 63647-5G 63647-25G
77472-332EA 415.09 USD
77472-332 77484-958 77495-514 77504-834
2,2,2-trifluoro-1-(2-(methoxymethoxy)-4,5-dimethylphenyl)ethanone ≥97%
2,2,2-trifluoro-1-(2-(methoxymethoxy)-4,5-dimethylphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₃F₃O₃
MW: 262.2250 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761470
CAS Number: 2901083-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR