(2-Chloro-6-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63605-1G 63605-500MG 63605-5G 63605-25G
77472-328EA 248.23 USD
77472-328 77484-954 77495-510 77504-830
(2-Chloro-6-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%
(2-Chloro-6-(methoxymethoxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁ClO₂S
MW: 218.7004 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761447
CAS Number: 2807448-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR