2-Chloro-3-fluoro-4-(methoxymethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

63581-1G 63581-500MG 63581-5G
77472-324EA 654.05 USD
77472-324 77484-950 77495-506
2-Chloro-3-fluoro-4-(methoxymethoxy)benzaldehyde ≥95%
2-Chloro-3-fluoro-4-(methoxymethoxy)benzaldehyde

Precaución: For research use only

Formula: C₉H₈ClFO₃
MW: 218.6094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761436
CAS Number: 2807441-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR