1-Bromo-2-chloro-3-fluoro-4-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

63540-1G 63540-500MG 63540-5G 63540-25G
77472-320EA 341.96 USD
77472-320 77484-946 77495-502 77504-824
1-Bromo-2-chloro-3-fluoro-4-(methoxymethoxy)benzene ≥95%
1-Bromo-2-chloro-3-fluoro-4-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇BrClFO₂
MW: 269.4953 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761411
CAS Number: 2807470-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR