2-Bromo-4-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63492-1G 63492-500MG 63492-5G
77472-316EA 473.8 USD
77472-316 77484-942 77495-498
2-Bromo-4-methoxy-5-methylbenzaldehyde ≥97%
2-Bromo-4-methoxy-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34761371
CAS Number: 2324882-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR