1-(6-Bromo-2-chloro-3-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

63465-1G 63465-500MG 63465-5G 63465-25G
77472-312EA 415.09 USD
77472-312 77484-938 77495-494 77504-818
1-(6-Bromo-2-chloro-3-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(6-Bromo-2-chloro-3-methylphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O
MW: 301.4876 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34814252
CAS Number: 2921833-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR