2',6'-Dichloro-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

63459-1G 63459-500MG 63459-5G
77472-308EA 476.89 USD
77472-308 77484-936 77495-488
2',6'-Dichloro-2,2,2-trifluoroacetophenone ≥95%
2',6'-Dichloro-2,2,2-trifluoroacetophenone

Precaución: For research use only

Formula: C₈H₃Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD08276246
CAS Number: 886510-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR